Learn More
Macherey Nagel Nanocolor Tube Test
Fast, easy-to-use, bar-coded, predosed test tubes for testing a variety of water and wastewater analysis parameters.
77.10 EUR - 108.00 EUR
Product Code | Brand | Analytes | Description | No. of Tests | Range | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Analytes | Description | No. of Tests | Range | Price | Quantity & Availability | ||
10702574
|
Macherey Nagel
985073 |
Sulfide 3, S2- | Seawater suitable after dilution (1 +3) | 20 Determinations | 0.05 - 3.00 mg/L |
92.50 EUR
1 test |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||
10151852
|
Macherey Nagel
985083 |
Total Nitrogen, TNb 22 | Not seawater suitable | 20 Determinations | 0.5 - 22.0 mg/L |
108.00 EUR
1 test |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||
11380625
|
Macherey Nagel
985008 |
Ammonium 100, NH4-N, NH4+ | Seawater suitable | 20 Determinations | 4 - 80 mg/L NH4-N, 5-100 mg/L NH4+ |
108.00 EUR
1 test |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||
11350635
|
Macherey Nagel
985049 |
Silver 3, Ag | Not seawater suitable | 20 Determinations | 0.20 - 3.00 mg/L |
77.10 EUR
1 test |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
15808775
|
Macherey Nagel
985062 |
Sulfate LR 2000, S42- | Not seawater suitable | 20 Determinations | 20 - 200 mg/L |
79.20 EUR
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||
Description
- Easy and fast measurements
- Using bar coded test tubes, the measurement is automatically performed after inserting the cuvette into the cuvette slot
- Contact with any dangerous chemicals is avoided because the measurement is performed directly in the test tube
- This guarantees maximum safety for the user
- Approved
- Reliable and comparable results
- The reaction basis of tests is based on internationally approved standard methods including DIN-, EN-, ISO-, EPA, and APHA
- Versatile
- The perfect test for every request
- A large number of tests with different ranges is available for all important parameters in water and wastewater analysis
- Precise
- Easy handling and highest accuracy
- All tests contain accurate predosed reagents that are ready to use
- Unique
- No mix up of tests
- Every test box is equipped with a color-coded label and can be clearly identified
Specifications
20 Determinations | |
Nanocolor™ PhotometricTube Test |