Learn More
Macherey Nagel Nanocolor Tube Test
Fast, easy-to-use, bar-coded, predosed test tubes for testing a variety of water and wastewater analysis parameters.
Brand:  Macherey Nagel 985062
Additional Details : CAS Number : 57-09-0 Weight : 0.34500kg
 View more versions of this product
Product Code. 15808775
  • 79.20 EUR / Each
Estimated Shipment: 12-07-2024
to see stock.
Add to basket
Description
Description
  • Easy and fast measurements
    • Using bar coded test tubes, the measurement is automatically performed after inserting the cuvette into the cuvette slot
    • Contact with any dangerous chemicals is avoided because the measurement is performed directly in the test tube
    • This guarantees maximum safety for the user
  • Approved
    • Reliable and comparable results
    • The reaction basis of tests is based on internationally approved standard methods including DIN-, EN-, ISO-, EPA, and APHA
  • Versatile
    • The perfect test for every request
    • A large number of tests with different ranges is available for all important parameters in water and wastewater analysis
  • Precise
    • Easy handling and highest accuracy
    • All tests contain accurate predosed reagents that are ready to use
  • Unique
    • No mix up of tests
    • Every test box is equipped with a color-coded label and can be clearly identified
Specifications
Specifications
Sulfate LR 2000, S42-
20 Determinations
By the addition of barium salts, sulfate forms hardly soluble Barium sulfate. The generated turbidity is measured photometrically at 585 nm.
Not seawater suitable
20 - 200 mg/L
Nanocolor™ PhotometricTube Test
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • NANOCOLOR Sulfate LR 200
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contains : barium chloride, hexadecyl-trimethylammonium bromide, EDTA-Na