Learn More
Macherey Nagel Nanocolor Tube Test
Fast, easy-to-use, bar-coded, predosed test tubes for testing a variety of water and wastewater analysis parameters.
Brand: Macherey Nagel 985083
Additional Details : Weight : 1.00000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
- Easy and fast measurements
- Using bar coded test tubes, the measurement is automatically performed after inserting the cuvette into the cuvette slot
- Contact with any dangerous chemicals is avoided because the measurement is performed directly in the test tube
- This guarantees maximum safety for the user
- Approved
- Reliable and comparable results
- The reaction basis of tests is based on internationally approved standard methods including DIN-, EN-, ISO-, EPA, and APHA
- Versatile
- The perfect test for every request
- A large number of tests with different ranges is available for all important parameters in water and wastewater analysis
- Precise
- Easy handling and highest accuracy
- All tests contain accurate predosed reagents that are ready to use
- Unique
- No mix up of tests
- Every test box is equipped with a color-coded label and can be clearly identified
Specifications
Total Nitrogen, TNb 22 | |
20 Determinations | |
Oxidative decomposition in the heating block with subsequent interference compensation and photometric determination with 2,6-dimethylphenol in sulphuric acid / phosphoric acid mixture |
Not seawater suitable | |
0.5 - 22.0 mg/L | |
Nanocolor™ PhotometricTube Test |
Safety and Handling
- NANOCOLOR total Nitrogen TNb 22
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Oxidising solid Category 2
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : 2 -propanol, o -phosphoric acid, sulfuric acid, sodium carbonate, potassium peroxydisulfate