Learn More
Macherey Nagel Nanocolor Tube Test
Fast, easy-to-use, bar-coded, predosed test tubes for testing a variety of water and wastewater analysis parameters.
Brand:  Macherey Nagel 985008
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11380625
  • 108.00 EUR / 1 test
In Stock
Description
  • Easy and fast measurements
    • Using bar coded test tubes, the measurement is automatically performed after inserting the cuvette into the cuvette slot
    • Contact with any dangerous chemicals is avoided because the measurement is performed directly in the test tube
    • This guarantees maximum safety for the user
  • Approved
    • Reliable and comparable results
    • The reaction basis of tests is based on internationally approved standard methods including DIN-, EN-, ISO-, EPA, and APHA
  • Versatile
    • The perfect test for every request
    • A large number of tests with different ranges is available for all important parameters in water and wastewater analysis
  • Precise
    • Easy handling and highest accuracy
    • All tests contain accurate predosed reagents that are ready to use
  • Unique
    • No mix up of tests
    • Every test box is equipped with a color-coded label and can be clearly identified
Specifications
Specifications
Ammonium 100, NH4-N, NH4+
20 Determinations
DIN method: At a pH value of about 12.6 ammonium ions react with hypochlorite and salicylate in the presence of sodium nitroprussiate as catalyst to form a blue indophenol.
Seawater suitable
4 - 80 mg/L NH4-N, 5-100 mg/L NH4+
Nanocolor™ PhotometricTube Test