Learn More
Macherey Nagel Nanocolor Tube Test
Fast, easy-to-use, bar-coded, predosed test tubes for testing a variety of water and wastewater analysis parameters.
Brand: Macherey Nagel 985008
Additional Details : Weight : 0.01000kg
Description
- Easy and fast measurements
- Using bar coded test tubes, the measurement is automatically performed after inserting the cuvette into the cuvette slot
- Contact with any dangerous chemicals is avoided because the measurement is performed directly in the test tube
- This guarantees maximum safety for the user
- Approved
- Reliable and comparable results
- The reaction basis of tests is based on internationally approved standard methods including DIN-, EN-, ISO-, EPA, and APHA
- Versatile
- The perfect test for every request
- A large number of tests with different ranges is available for all important parameters in water and wastewater analysis
- Precise
- Easy handling and highest accuracy
- All tests contain accurate predosed reagents that are ready to use
- Unique
- No mix up of tests
- Every test box is equipped with a color-coded label and can be clearly identified
Specifications
Ammonium 100, NH4-N, NH4+ | |
20 Determinations | |
DIN method: At a pH value of about 12.6 ammonium ions react with hypochlorite and salicylate in the presence of sodium nitroprussiate as catalyst to form a blue indophenol. |
Seawater suitable | |
4 - 80 mg/L NH4-N, 5-100 mg/L NH4+ | |
Nanocolor™ PhotometricTube Test |