Learn More
Macherey Nagel Nanocolor Tube Test
Fast, easy-to-use, bar-coded, predosed test tubes for testing a variety of water and wastewater analysis parameters.
Brand: Macherey Nagel 985073
Additional Details : Weight : 0.25000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
- Easy and fast measurements
- Using bar coded test tubes, the measurement is automatically performed after inserting the cuvette into the cuvette slot
- Contact with any dangerous chemicals is avoided because the measurement is performed directly in the test tube
- This guarantees maximum safety for the user
- Approved
- Reliable and comparable results
- The reaction basis of tests is based on internationally approved standard methods including DIN-, EN-, ISO-, EPA, and APHA
- Versatile
- The perfect test for every request
- A large number of tests with different ranges is available for all important parameters in water and wastewater analysis
- Precise
- Easy handling and highest accuracy
- All tests contain accurate predosed reagents that are ready to use
- Unique
- No mix up of tests
- Every test box is equipped with a color-coded label and can be clearly identified
Specifications
Sulfide 3, S2- | |
20 Determinations | |
DIN method: N,N-Dimethyl-1,4-phenylene diamine reacts with hydrogen sulphide to form a compound which changes to leucomethylene blue. This compound is oxidized to methylene blue using iron(III) ions (Reaction basis according to DIN 38405-D26/27) |
Seawater suitable after dilution (1 +3) | |
0.05 - 3.00 mg/L | |
Nanocolor™ PhotometricTube Test |