Learn More
Macherey Nagel Nanocolor Tube Test
Fast, easy-to-use, bar-coded, predosed test tubes for testing a variety of water and wastewater analysis parameters.
Brand:  Macherey Nagel 985049
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11350635
  • 77.10 EUR / 1 test
Estimated Shipment: 10-07-2024
to see stock.
Add to basket
Description
Description
  • Easy and fast measurements
    • Using bar coded test tubes, the measurement is automatically performed after inserting the cuvette into the cuvette slot
    • Contact with any dangerous chemicals is avoided because the measurement is performed directly in the test tube
    • This guarantees maximum safety for the user
  • Approved
    • Reliable and comparable results
    • The reaction basis of tests is based on internationally approved standard methods including DIN-, EN-, ISO-, EPA, and APHA
  • Versatile
    • The perfect test for every request
    • A large number of tests with different ranges is available for all important parameters in water and wastewater analysis
  • Precise
    • Easy handling and highest accuracy
    • All tests contain accurate predosed reagents that are ready to use
  • Unique
    • No mix up of tests
    • Every test box is equipped with a color-coded label and can be clearly identified
Specifications
Specifications
Silver 3, Ag
20 Determinations
Silver ions react with an indicator to form a blue dye. Insoluble silver compounds like silver bromide, silver chloride, silver iodide, silver cyanide or silverthiocyanate are not detected with this test. These compounds can be determined after pretreatment with NANOCOLOR™ NanOx Metal (Cat. No. 918 978).
Not seawater suitable
0.20 - 3.00 mg/L
Nanocolor™ PhotometricTube Test