missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylthioacetamide, 97%, Thermo Scientific Chemicals

CAS: 631-67-4 | C4H9NS | 103.183 g/mol

45.60 EUR - 155.00 EUR

Chemical Identifiers

CAS 631-67-4
Molecular Formula C4H9NS
Molecular Weight (g/mol) 103.183
MDL Number MFCD00022178
InChI Key LKNQXZAHNDFIQY-UHFFFAOYSA-N
Synonym n,n-dimethylthioacetamide, ethanethioamide, n,n-dimethyl, dimethylthioacetamide, dimethylthioacetamid, acetamide, n,n-dimethylthio, dimethylthioacetamid german, nn-dimethylthioacetamide, acmc-1b8ok, 4-04-00-00182 beilstein handbook reference
PubChem CID 12434
IUPAC Name N,N-dimethylethanethioamide
SMILES CC(=S)N(C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11410481
SDS View Documents
Thermo Scientific Alfa Aesar
B20020.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11420481
SDS View Documents
Thermo Scientific Alfa Aesar
B20020.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

N,N-Dimethylthioacetamide is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylthioacetamide is used as pharmaceutical intermediates.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylthioacetamide, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.