missing translation for 'onlineSavingsMsg'
Learn More
N,N-Dimethylthioacetamide, 97% Product Code.: 11420481

N,N-Dimethylthioacetamide, 97%

Product Code. 11420481
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11420481

Brand: Thermo Scientific Alfa Aesar B20020.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 631-67-4 | C4H9NS | 103.183 g/mol

N,N-Dimethylthioacetamide is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylthioacetamide is used as pharmaceutical intermediates.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 631-67-4
Molecular Formula C4H9NS
Molecular Weight (g/mol) 103.183
MDL Number MFCD00022178
InChI Key LKNQXZAHNDFIQY-UHFFFAOYSA-N
Synonym n,n-dimethylthioacetamide, ethanethioamide, n,n-dimethyl, dimethylthioacetamide, dimethylthioacetamid, acetamide, n,n-dimethylthio, dimethylthioacetamid german, nn-dimethylthioacetamide, acmc-1b8ok, 4-04-00-00182 beilstein handbook reference
PubChem CID 12434
IUPAC Name N,N-dimethylethanethioamide
SMILES CC(=S)N(C)C

Specifications

Melting Point 72°C to 75°C
Boiling Point 125°C to 127°C (22 mmHg)
Quantity 5 g
Formula Weight 103.19
Percent Purity 97%
Chemical Name or Material N,N-Dimethylthioacetamide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberE4200B Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20020.03,B20020.06
Lot Number10137937 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20020.03,B20020.06
Lot NumberH09X031 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20020.03,B20020.06
Lot NumberI17T032 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20020.03,B20020.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N-Dimethylthioacetamide, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.