Learn More
Fisherbrand™ Colored PTFE Conical Joint Clamp
Fisherbrand™ Colored PTFE Conical Joint Clamps for glass joints and can be used as a safe solution to clamping glass joints. The PTFE conical joint clamps include an encapsulated steel spring.
25.75 EUR - 42.50 EUR
Specifications
Quantity | 1 |
---|---|
Material | PTFE |
For Use With (Equipment) | Standard Taper Ground Glass Joints |
Description | Fisherbrand™ Colored PTFE Conical Joint Clamp |
Product Code | Brand | Color | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Size | Price | Quantity & Availability | ||||
16256031
|
Fisherbrand™
|
Green | Joint 10/30 |
27.15 EUR
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
16276031
|
Fisherbrand™
|
Purple | Joint 12/30 |
27.15 EUR
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
16296031
|
Fisherbrand™
|
Yellow | Joint 14/35 |
25.75 EUR
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
16216041
|
Fisherbrand™
|
Blue | Joint 19/38 |
27.95 EUR
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
16236041
|
Fisherbrand™
|
Green | Joint 24/40 |
28.45 EUR
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
16246041
|
Fisherbrand™
|
Red | Joint 29/42 |
30.25 EUR
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
16256041
|
Fisherbrand™
|
Orange | Joint 34/45 |
34.85 EUR
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
16266041
|
Fisherbrand™
|
Yellow | Joint 40/50 |
38.45 EUR
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
16286041
|
Fisherbrand™
|
Brown | Joint 45/50 |
42.50 EUR
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
Description
- Chemically inert and chemically resistant
- Temperature resistant, maintaining strength up to 280°C
- Fit stndard glass joints
- Can be adjusted by hand
Specifications
1 | |
PTFE | |
Standard Taper Ground Glass Joints | |
Fisherbrand™ Colored PTFE Conical Joint Clamp |