Learn More
Fisherbrand™ Colored PTFE Conical Joint Clamp
Fisherbrand™ Colored PTFE Conical Joint Clamps for glass joints and can be used as a safe solution to clamping glass joints. The PTFE conical joint clamps include an encapsulated steel spring.
Brand: Fisherbrand™
Additional Details : Weight : 0.01500kg
Description
- Chemically inert and chemically resistant
- Temperature resistant, maintaining strength up to 280°C
- Fit stndard glass joints
- Can be adjusted by hand
Specifications
1 | |
Standard Taper Ground Glass Joints | |
Joint 19/38 |
PTFE | |
Fisherbrand™ Colored PTFE Conical Joint Clamp | |
Blue |