missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Carbonate, Honeywell Fluka™
Puriss. p.a., ACS reagent
26.74 EUR - 205.17 EUR
Chemical Identifiers
CAS | 513-77-9 |
---|---|
Molecular Formula | CBaO3 |
Molecular Weight (g/mol) | 197.34 |
MDL Number | MFCD00003448 |
InChI Key | AYJRCSIUFZENHW-UHFFFAOYSA-L |
Synonym | barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p |
PubChem CID | 10563 |
SMILES | [Ba++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15681070
|
Honeywell-Fluka
11729-50G |
50 g | N/A | ||||||
15671070
|
Honeywell-Fluka
11729-250G |
250 g | N/A | ||||||
15661070
|
Honeywell-Fluka
11729-1KG |
1 kg |
205.17 EUR
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
513-77-9 | |
197.34 | |
AYJRCSIUFZENHW-UHFFFAOYSA-L | |
10563 |
CBaO3 | |
MFCD00003448 | |
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p | |
[Ba++].[O-]C([O-])=O |
Specifications
513-77-9 | |
BaCO3 | |
NONH for all modes of transport | |
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p | |
[Ba++].[O-]C([O-])=O | |
197.34 | |
197.34g/mol | |
Barium Carbonate |
CBaO3 | |
MFCD00003448 | |
7045119 | |
AYJRCSIUFZENHW-UHFFFAOYSA-L | |
barium(2+) carbonate | |
10563 | |
ACS/puriss. p.a. |
Safety and Handling
P301 + P312 + P330
H302
missing translation for 'einecsNumber' : 208-167-3
missing translation for 'rtecsNumber' : CQ8600000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title