missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Carbonate, Honeywell Fluka™
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 250 g |
Linear Formula | BaCO3 |
UN Number | NONH for all modes of transport |
Beilstein | 7045119 |
IUPAC Name | barium(2+);carbonate |
Formula Weight | 197.34g/mol |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Barium Carbonate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction