missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMPSTE24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | ZMPSTE24 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16870784
|
Proteintech
12847-1-AP-20UL |
20 μL |
140.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16860784
|
Proteintech
12847-1-AP-150UL |
150 μL |
399.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the peptidase M48A family. The encoded protein is a zinc metalloproteinase involved in the two step post-translational proteolytic cleavage of carboxy terminal residues of farnesylated prelamin A to form mature lamin A. Mutations in this gene have been associated with mandibuloacral dysplasia and restrictive dermopathy.Specifications
ZMPSTE24 | |
Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
O75844, Q80W54 | |
10269, 230709, 313564 | |
ZMPSTE24 Fusion Protein Ag3949 | |
Primary | |
-20°C | |
ZMPSTE24 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CAAX prenyl protease 1 homolog, FACE 1, FACE1, STE24, Ste24p, ZMPSTE24 | |
Zmpste24 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |