Learn More
ZMPSTE24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12847-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the peptidase M48A family. The encoded protein is a zinc metalloproteinase involved in the two step post-translational proteolytic cleavage of carboxy terminal residues of farnesylated prelamin A to form mature lamin A. Mutations in this gene have been associated with mandibuloacral dysplasia and restrictive dermopathy.Specifications
ZMPSTE24 | |
Polyclonal | |
Unconjugated | |
ZMPSTE24 | |
CAAX prenyl protease 1 homolog, FACE 1, FACE1, STE24, Ste24p, ZMPSTE24 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10269, 230709, 313564 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75844, Q80W54 | |
Zmpste24 | |
ZMPSTE24 Fusion Protein Ag3949 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |