missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tubulin beta class V Mouse anti-Canine, Human, Mouse, Clone: 1D7B9, Proteintech
Mouse Monoclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | Tubulin beta class V |
---|---|
Clone | 1D7B9 |
Concentration | 1 mg/mL |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16898165
|
Proteintech
66362-1-IG-20UL |
20 μL |
140.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16888165
|
Proteintech
66362-1-IG-150UL |
150 μL |
399.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain.Specifications
Tubulin beta class V | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HsT1601, TUBB 5, TUBB6, Tubulin beta 6 chain, Tubulin beta class V, tubulin, beta 6 | |
TUBB6 | |
IgG1 | |
Protein G | |
Antibody |
1D7B9 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
Unconjugated | |
Mouse | |
Canine, Human, Mouse | |
Q922F4, Q9BUF5 | |
480213, 67951, 84617 | |
Peptide | |
Primary | |
-20°C | |
TUBB6 |