missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tubulin beta class V Mouse anti-Canine, Human, Mouse, Clone: 1D7B9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66362-1-IG-150UL
This item is not returnable.
View return policy
Description
Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain.Specifications
Tubulin beta class V | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q922F4, Q9BUF5 | |
TUBB6 | |
Peptide | |
150 μL | |
Primary | |
Canine, Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1D7B9 | |
Unconjugated | |
TUBB6 | |
HsT1601, TUBB 5, TUBB6, Tubulin beta 6 chain, Tubulin beta class V, tubulin, beta 6 | |
Mouse | |
Protein G | |
RUO | |
480213, 67951, 84617 | |
-20°C | |
Liquid |