Learn More
DWK Life Sciences Kimble™ Kontes™ KIMAX™ Tooled Top Narrow Mouth Reservoir Bottle with Bottom Hose Outlet
Glass hose connection outlet is fused to the bottle
695.00 EUR - 2111.87 EUR
Specifications
For Use With (Application) | Designed to store and discharge liquids via a bottom hose outlet |
---|---|
Includes | Bottom hose outlet |
Material | Borosilicate Glass |
Mouth | Narrow |
Product Type | Reservoir Bottle |
Product Code | Brand | Capacity (English) | Height (English) | Stopper Number | Capacity (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (English) | Height (English) | Stopper Number | Capacity (Metric) | Price | Quantity & Availability | ||
11717969
|
DWK Life Sciences
KC14607250 |
8.45 oz. | 5.19 in. | 2 | 250 mL | ||||
11727969
|
DWK Life Sciences
KC14607500 |
16.91 oz. | 6.49 in. | 4 | 500 mL | ||||
11737969
|
DWK Life Sciences
KC146071000 |
33.81 oz. | 7.87 in. | 6 | 1000 mL | ||||
11757969
|
DWK Life Sciences
KC146075000 |
169.07 oz. | 12.59 in. | 10 | 5000 mL |
695.00 EUR
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
||
11747969
|
DWK Life Sciences
KC146072000 |
67.62 oz. | 9.84 in. | 6 | 2000 mL | ||||
11767969
|
DWK Life Sciences
KC1460710000 |
338.14 oz. | 15.74 in. | 12 | 10,000 mL | ||||
11777969
|
DWK Life Sciences
KC1460720000 |
676.28 oz. | 19.48 in. | 12 | 20 L | ||||
Description
- KimCote™ plastic coating for added safety in the event of breakage
- With white enamel marking spot
- Rubber stopper used to cap narrow mouth tooled top
- Glass conforms to ASTM™ E 438, Type I, Class A specifications
- Sizes 250mL and 500mL accept 5/16 in. I.D. flexible tubing
- Sizes 1000mL and larger accept 3/8 in. I.D. tubing
Specifications
Designed to store and discharge liquids via a bottom hose outlet | |
Borosilicate Glass | |
Reservoir Bottle |
Bottom hose outlet | |
Narrow |