Learn More
DWK Life Sciences Kimble™ Kontes™ KIMAX™ Tooled Top Narrow Mouth Reservoir Bottle with Bottom Hose Outlet
Product Code. 11777969
Click to view available options
Capacity (English):
8.45 oz.
16.91 oz.
33.81 oz.
67.62 oz.
169.07 oz.
338.14 oz.
676.28 oz.
Height (English):
5.19 in.
6.49 in.
7.87 in.
9.84 in.
12.59 in.
15.74 in.
19.48 in.
Stopper Number:
2
4
6
10
12
Capacity (Metric):
10,000 mL
1000 mL
20 L
2000 mL
250 mL
500 mL
5000 mL
Glass hose connection outlet is fused to the bottle
  • KimCote™ plastic coating for added safety in the event of breakage
  • With white enamel marking spot
  • Rubber stopper used to cap narrow mouth tooled top
  • Glass conforms to ASTM™ E 438, Type I, Class A specifications
  • Sizes 250mL and 500mL accept 5/16 in. I.D. flexible tubing
  • Sizes 1000mL and larger accept 3/8 in. I.D. tubing
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 20 L
Capacity (English) 676.28 oz.
Diameter (English) 11.41 in.
Diameter (English) Inner Neck 2.28 in.
Diameter (Metric) Inner Neck 58 mm
For Use With (Application) Designed to store and discharge liquids via a bottom hose outlet
Includes Bottom hose outlet
Material Borosilicate Glass
Stopper Number 12
Mouth Narrow
Height (Metric) 495 mm
Product Type Reservoir Bottle
Height (English) 19.48 in.
Diameter (Metric) 290 mm
Diameter (English) Inner Tubing 0.37 in.
Diameter (Metric) Inner Tubing 9.52 mm