missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Munktell Filtrak™ Grade MGA Binder-Free Micro-Glass Fiber Filters
For general laboratory filtration, clarification of buffer and reagent solutions.
Brand: Munktell Filtrak™ 410124
Additional Details : Weight : 0.04500kg
Description
- Made of high-purity borosilicate glass microfibers
- Biologically inert and resistant to most solvents and reagents with exception of hydrofluoric acid and highly concentrated alkali solutions
- Temperature resistant to about 500°C
Specifications
Disc | |
1.6 μm | |
MGA | |
55 mm |
Borosilicate Glass | |
General laboratory filtration, clarification of buffer and reagent solutions, corresponds to many international standards for air and water pollution monitoring. Foodstuff analysis, filtration of algae, bacteria cultures, proteins | |
0.23 mm |