missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Munktell Filtrak™ Grade MGA Binder-Free Micro-Glass Fiber Filters
For general laboratory filtration, clarification of buffer and reagent solutions.
44.00 EUR - 77.00 EUR
Specifications
Format | Disc |
---|---|
Material | Borosilicate Glass |
Particle Retention | 1.6 μm |
For Use With (Application) | General laboratory filtration, clarification of buffer and reagent solutions, corresponds to many international standards for air and water pollution monitoring. Foodstuff analysis, filtration of algae, bacteria cultures, proteins |
Grade | MGA |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
11377274
|
Munktell Filtrak™
410123 |
90 mm |
77.00 EUR
Pack of 100 |
Estimated Shipment: 19-04-2024
Log in to see stock. |
|||||
11357274
|
Munktell Filtrak™
410124 |
55 mm |
44.00 EUR
Pack of 100 |
In Stock
Log in to see stock. |
|||||
Description
- Made of high-purity borosilicate glass microfibers
- Biologically inert and resistant to most solvents and reagents with exception of hydrofluoric acid and highly concentrated alkali solutions
- Temperature resistant to about 500°C
Specifications
Disc | |
1.6 μm | |
MGA |
Borosilicate Glass | |
General laboratory filtration, clarification of buffer and reagent solutions, corresponds to many international standards for air and water pollution monitoring. Foodstuff analysis, filtration of algae, bacteria cultures, proteins | |
0.23 mm |