missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C21orf66 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | C21orf66 |
---|---|
Concentration | 0.58 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16830965
|
Proteintech
21357-1-AP-20UL |
20 μL |
140.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16820965
|
Proteintech
21357-1-AP-150UL |
150 μL |
399.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a protein that may bind to GC-rich DNA sequences, which suggests its involvement in the regulation of transcription. Alternative splicing of this gene results in multiple transcript variants.Specifications
C21orf66 | |
Immunofluorescence, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P58501, Q9Y5B6 | |
67367, 681004, 94104 | |
C21orf66 Fusion Protein Ag15789 | |
Primary | |
-20°C | |
PAXBP1 |
0.58 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BM 020, C21orf66, fSAP105, GCFC, GCFC1, PAXBP1 | |
Paxbp1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |