missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C21orf66 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21357-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that may bind to GC-rich DNA sequences, which suggests its involvement in the regulation of transcription. Alternative splicing of this gene results in multiple transcript variants.Specifications
C21orf66 | |
Polyclonal | |
Unconjugated | |
PAXBP1 | |
BM 020, C21orf66, fSAP105, GCFC, GCFC1, PAXBP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
67367, 681004, 94104 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.58 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P58501, Q9Y5B6 | |
Paxbp1 | |
C21orf66 Fusion Protein Ag15789 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |