Learn More
2-tert-Butyl-trans-vinylboronic acid, 97%, Thermo Scientific Chemicals
104.00 EUR - 323.00 EUR
Chemical Identifiers
CAS | 86595-37-1 |
---|---|
Molecular Formula | C6H13BO2 |
Molecular Weight (g/mol) | 127.978 |
MDL Number | MFCD01074624 |
InChI Key | CTIWKIMYFQSVBZ-SNAWJCMRSA-N |
Synonym | 3,3-dimethyl-1-butenylboronic acid, e-3,3-dimethylbut-1-en-1-yl boronic acid, 2-t-butyl-e-vinylboronic acid, 2-tert-butyl-e-vinylboronic acid, e-3,3-dimethylbut-1-enyl boronic acid, 1-butenyl-3,3-dimethyboronic acid, 1e-3,3-dimethylbut-1-en-1-ylboronic acid, boronic acid, b-3,3-dimethyl-1-buten-1-yl, 3,3-dimethylbut-1-enylboronic acid |
PubChem CID | 5706775 |
IUPAC Name | [(E)-3,3-dimethylbut-1-enyl]boronic acid |
SMILES | B(C=CC(C)(C)C)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15499377
|
Thermo Scientific Alfa Aesar
H52627.MD |
250 mg |
104.00 EUR
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15489377
|
Thermo Scientific Alfa Aesar
H52627.03 |
1 g |
323.00 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-tert-Butyl-trans-vinylboronic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-tert-Butyl-trans-vinylboronic acid is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
86595-37-1 | |
127.978 | |
CTIWKIMYFQSVBZ-SNAWJCMRSA-N | |
5706775 | |
B(C=CC(C)(C)C)(O)O |
C6H13BO2 | |
MFCD01074624 | |
3,3-dimethyl-1-butenylboronic acid, e-3,3-dimethylbut-1-en-1-yl boronic acid, 2-t-butyl-e-vinylboronic acid, 2-tert-butyl-e-vinylboronic acid, e-3,3-dimethylbut-1-enyl boronic acid, 1-butenyl-3,3-dimethyboronic acid, 1e-3,3-dimethylbut-1-en-1-ylboronic acid, boronic acid, b-3,3-dimethyl-1-buten-1-yl, 3,3-dimethylbut-1-enylboronic acid | |
[(E)-3,3-dimethylbut-1-enyl]boronic acid |
Specifications
86595-37-1 | |
C6H13BO2 | |
3,3-dimethyl-1-butenylboronic acid, e-3,3-dimethylbut-1-en-1-yl boronic acid, 2-t-butyl-e-vinylboronic acid, 2-tert-butyl-e-vinylboronic acid, e-3,3-dimethylbut-1-enyl boronic acid, 1-butenyl-3,3-dimethyboronic acid, 1e-3,3-dimethylbut-1-en-1-ylboronic acid, boronic acid, b-3,3-dimethyl-1-buten-1-yl, 3,3-dimethylbut-1-enylboronic acid | |
CTIWKIMYFQSVBZ-SNAWJCMRSA-N | |
[(E)-3,3-dimethylbut-1-enyl]boronic acid | |
5706775 | |
97% |
78°C to 80°C | |
MFCD01074624 | |
Slightly soluble in water. | |
B(C=CC(C)(C)C)(O)O | |
127.978 | |
127.98 | |
2-tert-Butyl-trans-vinylboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only