Learn More
2-tert-Butyl-trans-vinylboronic acid, 97%, Thermo Scientific Chemicals
Código de producto. 15489377
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones
2-tert-Butyl-trans-vinylboronic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-tert-Butyl-trans-vinylboronic acid is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Identificadores de productos químicos
CAS 86595-37-1
Molecular Formula C6H13BO2
Molecular Weight (g/mol) 127.978
MDL Number MFCD01074624
InChI Key CTIWKIMYFQSVBZ-SNAWJCMRSA-N
Synonym 3,3-dimethyl-1-butenylboronic acid, e-3,3-dimethylbut-1-en-1-yl boronic acid, 2-t-butyl-e-vinylboronic acid, 2-tert-butyl-e-vinylboronic acid, e-3,3-dimethylbut-1-enyl boronic acid, 1-butenyl-3,3-dimethyboronic acid, 1e-3,3-dimethylbut-1-en-1-ylboronic acid, boronic acid, b-3,3-dimethyl-1-buten-1-yl, 3,3-dimethylbut-1-enylboronic acid
PubChem CID 5706775
IUPAC Name [(E)-3,3-dimethylbut-1-enyl]boronic acid
SMILES B(C=CC(C)(C)C)(O)O
Especificaciones
Melting Point 78°C to 80°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 127.98
Percent Purity 97%
Chemical Name or Material 2-tert-Butyl-trans-vinylboronic acid