Learn More
2-Methyl-5-(trifluoromethoxy)benzoyl chloride, 97%, Thermo Scientific™
88.30 EUR - 350.00 EUR
Chemical Identifiers
CAS | 1261445-13-9 |
---|---|
Molecular Formula | C9H6ClF3O2 |
Molecular Weight (g/mol) | 238.59 |
MDL Number | MFCD18399758 |
InChI Key | DWHYIYNVPCGCJQ-UHFFFAOYSA-N |
Synonym | 2-methyl-5-trifluoromethoxy benzoyl chloride |
PubChem CID | 58342060 |
IUPAC Name | 2-methyl-5-(trifluoromethoxy)benzoyl chloride |
SMILES | CC1=C(C=C(C=C1)OC(F)(F)F)C(=O)Cl |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1261445-13-9 | |
238.59 | |
DWHYIYNVPCGCJQ-UHFFFAOYSA-N | |
58342060 | |
CC1=C(C=C(C=C1)OC(F)(F)F)C(=O)Cl |
C9H6ClF3O2 | |
MFCD18399758 | |
2-methyl-5-trifluoromethoxy benzoyl chloride | |
2-methyl-5-(trifluoromethoxy)benzoyl chloride |
Specifications
1261445-13-9 | |
MFCD18399758 | |
Moisture sensitive | |
DWHYIYNVPCGCJQ-UHFFFAOYSA-N | |
2-methyl-5-(trifluoromethoxy)benzoyl chloride | |
58342060 | |
97% | |
2-Methyl-5-(trifluoromethoxy)benzoyl chloride |
C9H6ClF3O2 | |
UN3265 | |
2-methyl-5-trifluoromethoxy benzoyl chloride | |
CC1=C(C=C(C=C1)OC(F)(F)F)C(=O)Cl | |
238.59 | |
238.59 | |
Liquid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures