Learn More
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione, 98%, Thermo Scientific Chemicals
107.00 EUR - 931.00 EUR
Chemical Identifiers
CAS | 7298-89-7 |
---|---|
Molecular Formula | C8H11ClO2 |
Molecular Weight (g/mol) | 174.624 |
MDL Number | MFCD00010502 |
InChI Key | VOBIHUAWDXUCPH-UHFFFAOYSA-N |
Synonym | chlorodimedone, 2-chlorodimedone, monochlorodimedone, 2-chloro-5,5-dimethyl-1,3-cyclohexanedione, 1,1-dimethyl-4-chloro-3,5-cyclohexanedione, 1,3-cyclohexanedione, 2-chloro-5,5-dimethyl, monochlorodimedon |
PubChem CID | 122278 |
IUPAC Name | 2-chloro-5,5-dimethylcyclohexane-1,3-dione |
SMILES | CC1(CC(=O)C(C(=O)C1)Cl)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15469115
|
Thermo Scientific Alfa Aesar
H51035.MD |
250 mg |
107.00 EUR
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11309266
|
Thermo Scientific Alfa Aesar
H51035.03 |
1 g |
268.00 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15407927
|
Thermo Scientific Alfa Aesar
H51035.06 |
5 g | |||||||
Description
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloro-5,5-dimethyl-1,3-cyclohexanedione is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
Chemical Identifiers
7298-89-7 | |
174.624 | |
VOBIHUAWDXUCPH-UHFFFAOYSA-N | |
122278 | |
CC1(CC(=O)C(C(=O)C1)Cl)C |
C8H11ClO2 | |
MFCD00010502 | |
chlorodimedone, 2-chlorodimedone, monochlorodimedone, 2-chloro-5,5-dimethyl-1,3-cyclohexanedione, 1,1-dimethyl-4-chloro-3,5-cyclohexanedione, 1,3-cyclohexanedione, 2-chloro-5,5-dimethyl, monochlorodimedon | |
2-chloro-5,5-dimethylcyclohexane-1,3-dione |
Specifications
7298-89-7 | |
C8H11ClO2 | |
2326749 | |
chlorodimedone, 2-chlorodimedone, monochlorodimedone, 2-chloro-5,5-dimethyl-1,3-cyclohexanedione, 1,1-dimethyl-4-chloro-3,5-cyclohexanedione, 1,3-cyclohexanedione, 2-chloro-5,5-dimethyl, monochlorodimedon | |
VOBIHUAWDXUCPH-UHFFFAOYSA-N | |
2-chloro-5,5-dimethylcyclohexane-1,3-dione | |
122278 | |
98% |
161°C to 164°C | |
MFCD00010502 | |
Moisture sensitive | |
Insoluble in water. | |
CC1(CC(=O)C(C(=O)C1)Cl)C | |
174.624 | |
174.62 | |
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only