missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5,5-dimethyl-1,3-cyclohexanedione, 98%, Thermo Scientific Chemicals

Product Code. 11309266
Click to view available options
Quantity:
250 mg
1 g
5 g

2-Chloro-5,5-dimethyl-1,3-cyclohexanedione is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7298-89-7
Molecular Formula C8H11ClO2
Molecular Weight (g/mol) 174.624
MDL Number MFCD00010502
InChI Key VOBIHUAWDXUCPH-UHFFFAOYSA-N
Synonym chlorodimedone, 2-chlorodimedone, monochlorodimedone, 2-chloro-5,5-dimethyl-1,3-cyclohexanedione, 1,1-dimethyl-4-chloro-3,5-cyclohexanedione, 1,3-cyclohexanedione, 2-chloro-5,5-dimethyl, monochlorodimedon
PubChem CID 122278
IUPAC Name 2-chloro-5,5-dimethylcyclohexane-1,3-dione
SMILES CC1(CC(=O)C(C(=O)C1)Cl)C

Specifications

Melting Point 161°C to 164°C
Quantity 1 g
Beilstein 2326749
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 174.62
Percent Purity 98%
Chemical Name or Material 2-Chloro-5,5-dimethyl-1,3-cyclohexanedione

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'