Learn More
Cytiva XK Column Superose™ 4 Fast Flow
Prepacked with Superose™ 4 FF, a highly cross-linked 4% agarose matrix that provides excellent physical stability and chromatographic qualities
Brand:  Cytiva 29268020
Product Code. 15844192
  • 4096.00 EUR / Each
Estimated Shipment: 18-02-2025
Description
  • Developed specifically to meet the high throughput demands of industrial process separations
  • Enables the rapid processing of large volumes which makes these resins an excellent choice for use at process-scale
  • Rigidity permits high flow rates which provides good resolution in a minimal amount of time
  • Low non-specific interactions and excellent recoveries
  • Meets industrial demands with security of supply in addition to comprehensive technical and regulatory support
TRUSTED_SUSTAINABILITY
Specifications
Specifications
90 μm
76 to 130 mL
16 mm
380 to 650 mm
2 to 12
Sepharose™ 4 Fast Flow
Stand-Alone Pump Chromatography Systems
Superose Pre-packed Column
Ion Exchange Chromatography
5 bar (72.5 psi)
1 Adaptor
XC Column
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Sepharose 4 Fast Flow, XK 16/70
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol