Learn More
DWK Life Sciences Wheaton™ Black Phenolic Cap with Rubber Liner
14B rubber-lined caps feature a nontoxic, white, styrene-butadiene rubber liner, ideal for cell culture work
Brand:  DWK Life Sciences W239306
 View more versions of this product
Product Code. 12485951
  • 145.00 EUR / Pack of 72
Only null left
Estimated Shipment: 03-11-2025
Specifications
Black
1000 Cs.
ISO 9001
Rubber
Phenolic Resin
48-400
Bottle
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Black Phenolic Cap with Rubber Liner > Screw cap size: 48mm-400