Learn More
DWK Life Sciences Wheaton™ Black Phenolic Cap with Rubber Liner
14B rubber-lined caps feature a nontoxic, white, styrene-butadiene rubber liner, ideal for cell culture work
Brand: DWK Life Sciences W239306
Description
Specifications
Black | |
1000 Cs. | |
ISO 9001 | |
Rubber |
Phenolic Resin | |
48-400 | |
Bottle |