Learn More
Cytiva Whatman™ Glass Microfiber Filter Paper Grade 6
Captures tiny particles without becoming weak. Whatman™ Glass Microfiber Filter Paper Grade 6 may also be used for determination of filterable substances and residue upon ignition or analysis of aggressive media.
64.20 EUR - 830.00 EUR
Specifications
Material | Borosilicate Glass |
---|---|
Particle Retention | <1 μm |
Filtration Speed | 40 s/100mL/in2 |
Flow Rate | Slow |
Grade | GF 6 |
Product Code | Brand | Diameter (Metric) | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Format | Price | Quantity & Availability | ||||
11385254
|
Cytiva
10370002 |
50 mm | Disc |
110.00 EUR
Pack of 200 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
11305264
|
Cytiva
10370004 |
70 mm | Disc |
127.00 EUR
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
11395254
|
Cytiva
10370003 |
55 mm | Disc |
99.80 EUR
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
11315264
|
Cytiva
10370005 |
90 mm | Disc |
177.00 EUR
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
11325264
|
Cytiva
10370006 |
110 mm | Disc |
287.00 EUR
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
11335264
|
Cytiva
10370007 |
125 mm | Disc |
302.00 EUR
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
11345264
|
Cytiva
10370008 |
150 mm | Disc |
414.00 EUR
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
11355264
|
Cytiva
10370010 |
185 mm | Disc |
463.00 EUR
Pack of 100 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
11375264
|
Cytiva
10370012 |
240 mm | Disc |
610.00 EUR
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
11365264
|
Cytiva
10370011 |
200 mm | Disc |
540.00 EUR
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
11305274
|
Cytiva
10370020 |
100 mm | Disc |
280.00 EUR
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
11395264
|
Cytiva
10370019 |
47 mm | Disc |
97.65 EUR
Pack of 200 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
11385264
|
Cytiva
10370018 |
25 mm | Disc |
64.20 EUR
Pack of 200 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
11315274
|
Cytiva
10370050 |
620 mm | Sheet |
830.00 EUR
Pack of 100 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
Description
Suitable for water and wastewater analysis, with extremely fine particle retention and added strength.- Sub micron filter retention efficiency to capture extremely small particles
- Inorganic binder for increased strength
- Temperature-resistant up to 500°C
- Can be used as a pre-filter for dense samples
Specifications
Borosilicate Glass | |
40 s/100mL/in2 | |
GF 6 |
<1 μm | |
Slow | |
350 μm |