Learn More
CleanSpace™ PAPR Half Mask
Half masks clip quickly onto PAPR power units and easily unclip for cleaning. Specifically designed to be used with the CleanSpace™ 2, Ultra and EX PAPR systems.
Brand: CleanSpace™ PAF-1010
Description
- Compatible with ALL CleanSpace™PAPR power units
- Used with the revolutionary CleanSpace™PAPR: lightweight, no hoses or belts
- Available in 3 sizes to facilitate a secure seal and a comfortable fit
- Materials: Soft silicone cushion, nylon clips and polycarbonate (exhalation valve)
- Easy removal for cleaning
- Transparent and facilitates communication
- Clip quickly onto the power units and are easily unclipped for cleaning
- Soft silicone that readily moulds to the face and does not cause allergic dermatitis
Specifications
AS/NZS1716: 2012 | |
EN 12942 | |
Medium | |
Mining, welding, manufacturing, smelting, construction, recycling plants, emergency services, agriculture, processing plants, grinding | |
IP66/67 | |
Store at -10°C to 55°C | |
125 g |
EN 12942 | |
PAPR Half Mask | |
PAPR half mask only | |
PAF-1034, PAF-2034 | |
PAF-1010 | |
170 x 130 x 70 mm |