Learn More
Invitrogen™ Lipofectamine™ RNAiMAX Transfection Reagent
Provides highest transfection efficiencies on widest variety of cell types for siRNA-mediated gene knockdown experiments
Brand: Invitrogen™ 13778030
345.80 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Description
Lipofectamine™ RNAiMAX Transfection Reagent provides the highest transfection efficiencies on the widest variety of cell types for siRNA-mediated gene knockdown experiments. Lipofectamine™ RNAiMAX is a proprietary RNAi-specific cationic lipid formulation designed specifically for the delivery of siRNA and miRNA into all cell types.With Lipofectamine™ RNAiMAX Transfection Reagent you will get:
High knockdown in a wide range of cells
Lipofectamine™ RNAiMAX Transfection Reagent transfects a wide range of cell types (see figure). For gene silencing, Lipofectamine™ RNAiMAX Transfection Reagent's high-efficiency transfections lead to the high levels of gene knockdown needed to achieve convincing results.
Simple, high-throughput–ready transfections
Simply mix Lipofectamine™ RNAiMAX Transfection Reagent with siRNA, add to your cells, incubate, and measure gene knockdown. The simplicity and speed combined with high transfection efficiency make Lipofectamine™ RNAiMAX Transfection Reagent ideal for high-throughput siRNA transfections. Transfection conditions can be easily established for automated or robotic systems used in such applications.
Find an optimized Lipofectamine™ RNAiMAX transfection protocol for your cell line.
Order Info
Shipping Condition: Qualified for Ambient or Wet Ice
Specifications
Each unit contains one vial of Lipofectamine™ RNAiMAX™ Reagent (0.3 ml). Store at 2–8°C. |
|
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks | |
Lipid-based Transfection | |
Sustainable packaging | |
Lipofectamine™ | |
0.3 mL | |
Approved for shipment at Room Temperature or on Wet Ice |
Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells | |
siRNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR), RNAi, miRNA | |
Transfection | |
High-throughput Compatible | |
Transfection Reagent | |
Yes |
For Research Use Only. Not for use in diagnostic procedures.