Learn More
Cole-Parmer™ VapLock™ Solvent Waste Kits for Jars and Pails
VapLock™ Solvent Waste Kits for Jars and Pails collect effluent from multiple sources while minimizing harmful solvent exhaust
Brand:  Cole-Parmer™ WAK100X-10
 View more versions of this product
Product Code. 15854534
  • 861.00 EUR / Each
Estimated Shipment: 06-10-2025
Description
Various cap sizes available to fit your solvent container
Services
None
Specifications
Specifications
Solvent Safety
PTFE
1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F)
Solvent-Compatible Containers
100-415
Solvent Waste Kit
Documents
One moment while we fetch your results.
Product Title
Cole-Parmer™ VapLock™ Solvent Waste Kits for Jars and Pails > PTFE, 100-415, 1x1/2 in NPTF, 3x1/4 in NPTF, 6x1/4 in-28 UNFF; 1/ea