Learn More
Trifluoroacetic acid-d, for NMR, 99% atom D, Thermo Scientific Chemicals
CAS: 599-00-8 | C2HF3O2 | 115.029 g/mol
34.85 EUR - 416.00 EUR
Chemical Identifiers
CAS | 599-00-8 |
---|---|
Molecular Formula | C2HF3O2 |
Molecular Weight (g/mol) | 115.029 |
MDL Number | MFCD00044563 |
InChI Key | DTQVDTLACAAQTR-DYCDLGHISA-N |
PubChem CID | 71502 |
IUPAC Name | deuterio 2,2,2-trifluoroacetate |
SMILES | C(=O)(C(F)(F)F)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10347870
|
Thermo Scientific Acros
166400050 |
5 mL | Glass Bottle |
34.85 EUR
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10285590
|
Thermo Scientific Acros
166400100 |
10 mL | Glass Bottle |
56.10 EUR
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10184270
|
Thermo Scientific
166400250 |
25 mL | Glass Bottle |
111.00 EUR
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10311384
|
Thermo Scientific Acros
166401000 |
100 mL | Glass Bottle |
416.00 EUR
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMRChemical Identifiers
599-00-8 | |
115.029 | |
DTQVDTLACAAQTR-DYCDLGHISA-N | |
deuterio 2,2,2-trifluoroacetate |
C2HF3O2 | |
MFCD00044563 | |
71502 | |
C(=O)(C(F)(F)F)O |
Specifications
Liquid | |
1.5000g/mL | |
0.1% max. (Coulometric) | |
-15.0°C | |
C2HF3O2 | |
MFCD00044563 | |
DTQVDTLACAAQTR-DYCDLGHISA-N | |
deuterio 2,2,2-trifluoroacetate | |
71502 | |
NMR |
Colorless | |
98.5% min. | |
599-00-8 | |
75.0°C | |
CF3CO2D | |
trifluoroacetic acid-d, deuterotrifluoroacetic acid, acetic acid-d, trifluoro, acetic acid-d,2,2,2-trifluoro, acetic acid-d, 2,2,2-trifluoro, trifluoroacetic acid-d tfa-d, trifluoroacetic acid-d, 99.5 atom % d, tfa-d, cf3cood, trifluoroacetic 2h acid | |
C(=O)(C(F)(F)F)O | |
115.029 | |
115.03 | |
Trifluoroacetic acid-d, 99 atom%D |
Safety and Handling
GHS H Statement:
Harmful to aquatic life with long lasting effects.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Trifluoroacetic acid-d
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H332-Harmful if inhaled.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.