Learn More
Triethylamine solution for Ion Chromatography 1g/L H2O matrix, Fisher Chemical™ Product Code.: 12912225
Triethylamine solution for Ion Chromatography 1g/L H2O matrix, Fisher Chemical™
BIOPHARMA
Product Code. 12912225
250 mL
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Product Code. 12912225
Brand: Fisher Chemical A 051 250
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Quantity 250 mL
Solution Contains H2O
Concentration 1 g/L
Product Content Correction
Product Title
Triethylamine solution for Ion Chromatography 1g/L H2O matrix, Fisher Chemical™ >