Learn More
SCHOTT AG Lighting and Imaging™ Transmitted Light Stage
The Transmitted Light Stage provides extreme homogeneous illumination for all kinds of transparent specimens.
Brand: SCHOTT AG Lighting and Imaging™ 600400
Additional Details : Weight : 2.00000kg
Description
- EasyLED Transmitted light system.
- Diameter of 84 mm.
- Robust metal housing and a SCHOTT Opalika™ surface.
- Well-designed heat sink maximizes light density up to 12,000 cd/m2.
- Long lifetime of at least 50,000 hours.
- Power supply 100-240 V included.
- Compact and ergonomical.
- Integrated controller.
- Scratch resistant surface.
- Wide range power supply.
- International clip plug system.
Specifications
Transmitted Light Stage | |
84 mm | |
Power Supply | |
SCHOTT Opalika™ | |
84 mm |
3.3 in. | |
Stereo Microscopy illumination | |
50,000 Hours | |
100/240 V |