Learn More
Epredia™ TPX Single Sample Chamber, Caps and Filter Cards
For use with disposable white filter cards for sample volumes up to 0.5 mL and for use with disposable brown filter cards for sample volumes of 0.4 mL or less.
53.00 EUR - 665.00 EUR
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11922355
|
Epredia™
5991022 |
White Filter Cards |
113.00 EUR
Pack of 200 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
12096669
|
Epredia™
A78710018 |
TPX Sample Chamber |
665.00 EUR
Pack of 12 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
12066689
|
Epredia™
5991023 |
Brown Filter Cards |
53.00 EUR
Pack of 200 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
Description
TPX Single Sample Chamber
- Reusable
- Autoclavable
- Highly absorbent disposable filter cards for use with TPX chamber
- Not for use with Cytofunnel™ disposable sample chambers