Learn More
Sonics Materials™ Titanium Alloy Replaceable Tip
Robust reliable replacement tip for probe that is fabricatedfrom titanium alloy Ti-6Al-4V and is autoclavable.
Brand:  Sonics Materials™ 630-0407
 View more versions of this product
Product Code. 10374882
  • 65.70 EUR / Each
Estimated Shipment: 24-06-2025
Description
Replaceable tips are fabricated from titanium alloy Ti-6Al-4V and are autoclavable.
Specifications
Specifications
Replaceable Tip
19 mm
Titanium Alloy
1 Ea.
Documents
One moment while we fetch your results.
Product Title
Sonics Materials™ Titanium Alloy Replaceable Tip > Diameter: 19mm