Learn More
Codan™ Three-part Syringes
Use Codan™ Three-part Syringes to accommodate a variety of medical needs including: catheterization, hypodermic ear treatments, and more. The syringes are latex-free and contain no rubber or silicone. The plastic plunger has a smooth draw and a positive safety stop to prevent accidental spill
Brand: Codan™ 62.8604
This item is not returnable.
View return policy
Description
- Sterile
- Graduations
- Disposable
- Latex-free
Specifications
Autoclavable | |
Dual scale; Silicone seal | |
Single-use | |
100 mL | |
Catheter | |
Sterile |
Polypropylene | |
Catheter-tip | |
5 mL | |
Catheter | |
Yes |