Learn More
Thermo Scientific™ Nalgene™ Replacement Closures for Economy Wash Bottles
Click to view available options
Neck Finish:
24-400
28-400
Description
- Closure and stem molded in one piece for leakproof service
- Tip can be cut back to provide increased flow
- Tight fit of draw tube and closure prevents separation when squeezing bottle
- Leakproof†
Specifications
Specifications
Material | Polypropylene |
Neck Finish | 28-400 |
For Use With (Equipment) | Wash bottle |
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Replacement Closures for Economy Wash Bottles >
Spot an opportunity for improvement?Share a Content Correction