Learn More
Thermo Scientific™ High Purity Single Stage Brass Gas Regulators
Recommended for applications where precise flow control is not critical
940.00 EUR
Specifications
Range | 0 to 200 psig |
---|
Includes: Diffusion-resistant, diaphragm packless outlet valve to maintain system purity
Product Code | Brand | Range | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Type | Price | Quantity & Availability | ||||
10351505
|
Thermo Scientific™
60180-901 |
0 to 200 psig | CGA 580 with Triple Filter |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10280005
|
Thermo Scientific™
60180-909 |
0 to 200 psig | CGA 320 with Triple Filter |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- For non-corrosive analytical and process applications
- Includes a built-in Smart Purifier Base and Cartridge ensuring 99.99999% gas purity
- Specially designed, convoluted, stainless steel diaphragm provides good regulating performance and maximum purity integrity without the need for a soft seal
- Capable of withstanding an internal vacuum
- All regulators supplied with 1/8 in. stainless steel output adaptor
- Built in check valve allows for cylinder change without contaminating the gas line
Specifications
0 to 200 psig |