Learn More
Thermo Scientific™ Adhesive Seal Applicator
Make sealing PCR and qPCR plates or tube strips easy
Brand: Thermo Scientific™ AB1391
Description
Adhesive PCR Plate Seals are a reliable method of sealing plates for PCR applications. The high integrity sealing materials give the best protection against evaporation for high efficiency PCR.
Thermo Scientific Sealing Accessories feature the Cap-IT Tool, which helps you add domed caps to tube strips and plates and helps seal multiple tubes at the same time. The Adhesive Seal Applicator helps you effectively seal PCR and qPCR plates and storage plates.
Highlights
Cap-It Tool
- Designed to assist in applying domed caps to tube strips and plates
- It minimizes the pressure required to seal the caps effectively
- Enables multiple tubes to be sealed simultaneously
- Helps ensure effective and consistent sealing of PCR/qPCR plates or storage plates with any adhesive seal
Hazardous: No
Storage Condition: Ambient
Specifications
Polyethylene | |
1 Unit | |
Film, Seal, or Applicator | |
96/384-well Plate |