Learn More
Sampling Systems™ SteriWare™ LiquiDip Mono
Sampling Systems™ SteriWare™ LiquiDip Mono is ideal for taking cross sectional or point samples of liquids.
Brand: Sampling Systems™ 8920H-450S
This item is not returnable.
View return policy
Description
- Assembled and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
- BSE/TSE free
- Suitable for taking cross sectional samples
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Available in 2 different lengths
- Push the sampler into the product, ensuring the sampler is in the open position
- Push the sampler against the base of the container to close the valve
- Remove the sampler and pour out the liquid into a suitable container
- No cross contamination
- No cleaning required
Specifications
Liquid Sampler | |
Valve at one end | |
Single-use | |
21 mm |
HDPE | |
80 mL | |
Sterile | |
450 mm |