Learn More
Sampling Systems™ SteriWare™ Disposable Jumbo Pipette
BIOPHARMA
Sampling Systems™ SteriWare™ Disposable Jumbo Pipette is both quick and easy to use and is ideal for taking samples from low viscosity liquids.
Brand:  Sampling Systems™ 8051H-450S
Product Code. 15468163
  • 258.00 EUR / Pack of 50
Estimated Shipment: 04-06-2025
Description
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 & EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Highly visible color coded handle
  • Press thumb onto blue end of the tube until lowered to the correct sampling depth
  • At the required depth raise thumb until levels equalize
  • Press the thumb onto tube to seal in the sample
  • No cross contamination
  • No cleaning required
Specifications
Specifications
HDPE
80 mL
21 mm
Individually Bagged
Sterile
450 mm
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number120837 Certificate TypeCertificate of Compliance Date09/04/2025 Catalog Number8051H-450S
Lot Number110608 Certificate TypeCertificate of Compliance Date28/03/2024 Catalog Number8051H-450S
Lot Number110607 Certificate TypeCertificate of Compliance Date28/11/2023 Catalog Number8051H-450S
Lot Number118439 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number8051H-450S
Product Title
Sampling Systems™ SteriWare™ Disposable Jumbo Pipette > LengthMetric: 450mm