Learn More
Bel-Art™ Sterileware™ Flat Bottom Sampling Scoop
Flat bottom sampling scoops are used by pharmaceutical, food, cosmetic and chemical companies for general production or laboratory work.
Brand:  Bel-Art™ F36911-0250
 View more versions of this product
Product Code. 16652814
  • 357.92 EUR / Pack of 100
Only null left
Estimated Shipment: 08-08-2025
Description
  • Flat bottom allows for use as a weigh boat
  • Approximate volumes can be measured by removing excess powder
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High impact polystyrene (HIPS) scoops heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10-6)
  • Ready for one time use and disposal
  • Lot stamped for accurate tracing
  • Shipped with a Certificate of Processing for Sterilization
Specifications
Specifications
Scoop
Not Autoclavable
8 oz.
Single-use
100/Pk.
3.75 in.
High-impact Polystyrene
Flat
250 mL
Sterile
Bowl: 14.3 x 8.0 x 4.2 cm (5.625 x 3 .125 x 1.625 in.)
9.5 cm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bel-Art™ Sterileware™ Flat Bottom Sampling Scoop > 250 mL