Learn More
Kappes Systeme™ Standing Aids
POA - 126.30 EUR
Specifications
Adjustable (Height) | 550 to 900 mm |
---|---|
With Backrest | Yes |
Product Type | Standing Aid |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
12602075
|
Kappes Systeme™
8101.00.1001 |
Silver |
POA
Each |
N/A | |||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
10474832
|
Kappes Systeme™
8101.00.1014 |
Traffic Red |
126.30 EUR
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
Specifications
550 to 900 mm | |
Yes | |
Standing Aid |