Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML F1, NVLAP
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML F1 Tolerances
Brand:  Ohaus™ 80780352
975.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16602452
  • 1000.00 EUR / Each
Estimated Shipment: 18-06-2025
Description
  • Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
  • NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NVLAP Accredited Certificate
13 kg
50 mg
Troemner Alloy 8
10 kg
Protective Case
Documents
One moment while we fetch your results.
Product Title
Ohaus™ Stainless Steel Calibration Weights - OIML F1, NVLAP > 10 kg