Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML E1, NVLAP
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML E1 Tolerances
482.00 EUR - 5915.00 EUR
Specifications
Certifications/Compliance | NVLAP Accredited Certificate |
---|---|
Includes | Protective Case |
Product Code | Brand | Mass | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | ||||||
16652292
|
Ohaus™
30390941 |
1 g |
482.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
16642292
|
Ohaus™
30390940 |
2 g |
493.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
16632292
|
Ohaus™
30390939 |
5 g |
503.00 EUR
Each |
In Stock
Log in to see stock. |
||||||
16622292
|
Ohaus™
30390938 |
10 g |
513.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
16612292
|
Ohaus™
30390937 |
20 g |
530.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
16602292
|
Ohaus™
30390936 |
50 g |
570.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
16692282
|
Ohaus™
30390935 |
100 g |
611.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
16682282
|
Ohaus™
30390934 |
200 g |
667.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
16672282
|
Ohaus™
30390933 |
500 g |
820.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
16662282
|
Ohaus™
30390932 |
1 kg |
1045.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
16652282
|
Ohaus™
30390931 |
2 kg |
1805.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
16642282
|
Ohaus™
30390930 |
5 kg |
2510.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
16632282
|
Ohaus™
30390929 |
10 kg |
3290.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
16622282
|
Ohaus™
30390928 |
20 kg |
5915.00 EUR
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
||||||
Description
- OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
- Weights in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
- NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
Specifications
NVLAP Accredited Certificate | |
Protective Case |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Ohaus™ Stainless Steel Calibration Weights - OIML E1, NVLAP