Learn More
Memmert™ Stainless Steel Bottom Drip Tray for UF160, 15 mm
Non-perforated bottom drip trays constructed of stainless steel.
Brand:  Memmert™ UF160B04359
Product Code. 17539005
  • 200.00 EUR / Each
Only null left
Estimated Shipment: 21-07-2025
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
UF160 Oven
Bottom Drip Tray
Stainless Steel
Rim: 15 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Stainless Steel Bottom Drip Tray for UF160, 15 mm > For Use With: UF160