Learn More
Bochem™ Stainless Steel 18/10 Pharmacy Scoops
116.00 EUR - 127.00 EUR
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10575685
|
Bochem™
8830 |
220 mm |
116.00 EUR
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
10288054
|
Bochem™
8831 |
280 mm |
127.00 EUR
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
Specifications
Stainless Steel (18/10) | |
Pharmacy Scoop |